ADV428292942 AChemBlock

2-(3-Aminooxetan-3-yl)ethanol hemioxalate

  • MDL number MFCD27988028
  • Cas Number : 1523618-22-5
  • Molecular Weight : 324.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-3-aminooxetan-3-yl-ethanol-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6613
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart