ADV428292947 AChemBlock

(2S,4S)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxy)pyrrolidine-2-carboxylic acid

  • MDL number MFCD22415192
  • Cas Number : 464193-92-8
  • Molecular Weight : 409.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-pyrrolidinedicarboxylic-acid-4-1-1-dimethylethoxy-1-9h-fluoren-9-ylmethyl-ester-2s-4s.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5814
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart