ADV428292955 AChemBlock

4-(3-(2H-1,2,3-Triazo-2-yl)phenylamino)-2-((1R,2S)-2-aminocyclohexylamino) pyrimidine-5-carboxamide acetate

  • MDL number MFCD23105961
  • Cas Number : 1370261-96-3
  • Molecular Weight : 453.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-3-2h-1-2-3-triazo-2-yl-phenylamino-2-1r-2s-2-aminocyclohexylamino-pyrimidine-5-carboxamide-acetate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6536
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart