ADV428292972 AChemBlock

4-Methyl-1H-pyrazol-5-amine

  • Cas Number : 64781-79-9
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methyl-1H-pyrazol-5-amine
Inchi Code 1S/C4H7N3/c1-3-2-6-7-4(3)5/h2H,1H3,(H3,5,6,7)
Inchi Key KIHDRFDQWVMKES-UHFFFAOYSA-N

Description

Synonym 4-methyl-1h-pyrazol-5-amine
Purity 0.97
Physical Form yellow liquid
Shipping Temperature Store at 0-8
Country of Origin CN
Product Link https://www.achemblock.com/4-methyl-1h-pyrazol-5-amine.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/ac_sds?Catalog=F-4795
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart