ADV428292980 AChemBlock

N-Methyl-N-(oxetan-3-yl)azetidin-3-amine dihydrochloride

  • MDL number MFCD22581532
  • Cas Number : 1403767-34-9
  • Molecular Weight : 215.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-n-oxetan-3-yl-azetidin-3-amine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6733
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart