ADV428292985 AChemBlock

2-(1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)ethanamine

  • MDL number MFCD16083241
  • Cas Number : 1247501-81-0
  • Molecular Weight : 177.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-1-1-dioxidotetrahydro-2h-thiopyran-4-yl-ethanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6849
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart