ADV428292986 AChemBlock

1-Boc-5-bromo-3-(chloromethyl)-7-azaindole

  • MDL number MFCD17015935
  • Cas Number : 1234616-42-2
  • Molecular Weight : 345.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-5-bromo-3-chloromethyl-7-azaindole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2339
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart