ADV428292997 AChemBlock

rel-(1R,2S)-2-(methoxycarbonyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD28098189
  • Cas Number : 31420-52-7
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1r-2s-rel-1-2-cyclobutanedicarboxylic-acid-1-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7039
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart