ADV428293002 AChemBlock

4-Aminomethylpyrimidine dihydrochloride

  • MDL number MFCD07781107
  • Cas Number : 618446-08-5
  • Molecular Weight : 182.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethylpyrimidine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2474
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart