ADV428293006 AChemBlock

Ethyl-(1-methyl-azetidin-3-yl)-amine

  • MDL number MFCD24335256
  • Cas Number : 1434128-51-4
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-methyl-azetidin-3-yl-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7106
pictograms GHS07
hstatements H302;H313;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart