ADV428293008 AChemBlock

trans-tert-Butyl 3-hydroxy-3-methylcyclobutylcarbamate

  • MDL number MFCD20926193
  • Cas Number : 1363382-14-2
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-tert-butyl-3-hydroxy-3-methylcyclobutylcarbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7184
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart