ADV428293010 AChemBlock

Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride

  • MDL number MFCD27976569
  • Cas Number : 147927-61-5
  • Molecular Weight : 171.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/bicyclo-1-1-1-pentane-1-3-diamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7234
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart