ADV428293025 AChemBlock

6-Oxa-3-azabicyclo[3.1.1]heptane tosylate

  • MDL number MFCD27665413
  • Cas Number : 1339953-58-0
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-oxa-3-azabicyclo-3-1-1-heptane-tosylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6636-1
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart