ADV428293027 AChemBlock

5-Boc-2,5-Diazaspiro[3.5]nonane hemioxalate

  • MDL number MFCD26959176
  • Cas Number : 1523571-07-4
  • Molecular Weight : 542.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-boc-2-5-diazaspiro-3-5-nonane-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4164-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart