ADV428293037 AChemBlock

4-Aminomethylpyridazine dihydrochloride

  • MDL number MFCD27756609
  • Cas Number : 1028615-75-9
  • Molecular Weight : 182.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form orange solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethylpyridazine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3221-01
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart