ADV428293039 AChemBlock

4-(aminomethyl)tetrahydro-2H-thiopyran 1,1-dioxide hydrochloride

  • MDL number MFCD13193865
  • Cas Number : 1107645-98-6
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-1-dioxotetrahydro-2h-thiopyran-4-yl-methyl-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2867-01
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart