ADV428293040 AChemBlock

N,N-Dimethyl-3-azetidinecarboxamide hydrochloride

  • MDL number MFCD10565797
  • Cas Number : 927390-60-1
  • Molecular Weight : 164.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethyl-3-azetidinecarboxamide-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3421-01
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart