ADV428293043 AChemBlock

7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD03094742
  • Cas Number : 220247-73-4
  • Molecular Weight : 248.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1-2-3-4-tetrahydroisoquinoline-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6480-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart