ADV428293045 AChemBlock

tert-Butyl 2-cyanopiperazine-1-carboxylate hemioxalate

  • MDL number MFCD27986914
  • Cas Number : 1523617-98-2
  • Molecular Weight : 512.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-cyanopiperazine-1-carboxylate-hemioxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4199-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart