ADV428293046 AChemBlock

3-Aminopyrazolo[1,5-a]pyrimidine TFA

  • MDL number MFCD22421658
  • Cas Number : 1400764-19-3
  • Molecular Weight : 248.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-aminopyrazolo-1-5-a-pyrimidine-tfa.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5337-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart