ADV428293049 AChemBlock

Methyl 2-methyltetrahydrofuran-2-carboxylate

  • MDL number MFCD18651757
  • Cas Number : 1218915-91-3
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-methyltetrahydrofuran-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8024
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart