ADV428293078 AChemBlock

5-Hydroxy-1-naphthoic acid

  • MDL number MFCD11870759
  • Cas Number : 2437-16-3
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-hydroxy-1-naphthoic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8058
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart