ADV428293084 AChemBlock

(2R,4R)-4-fluoropyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD27986959
  • Cas Number : 1523541-82-3
  • Molecular Weight : 169.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-4r-4-fluoropyrrolidine-2-carboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=H-8063-01
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart