ADV428293091 AChemBlock

(R)-1-Methylpyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD27987988
  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form light-yellow liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/r-1-methylpyrrolidine-3-carboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6130-01
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart