ADV428293092 AChemBlock

4-Amino-1-ethyl-2(1H)-pyridinone hydrochloride

  • MDL number MFCD27987170
  • Cas Number : 1523618-09-8
  • Molecular Weight : 174.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-amino-1-ethyl-2-1h-pyridinone-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-5850-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart