ADV428293093 AChemBlock

cis-3-Hydroxy-3-methylcyclobutylamine hydrochloride

  • MDL number MFCD23105823
  • Cas Number : 1523606-23-6
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-3-hydroxy-3-methylcyclobutylamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-6305-01
pictograms GHS07
hstatements H302;H313;H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart