ADV428293188 AChemBlock

1-(Bromomethyl)cyclohexene

  • MDL number MFCD17015937
  • Cas Number : 37677-17-1
  • Molecular Weight : 175.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-bromomethyl-cyclohexene.html
Availability details for
Quantity
  • Availability
  • Add to Cart