ADV428293191 AChemBlock

3,4-Dihydro-1H-isochromen-4-amine hydrochloride

  • MDL number MFCD08448171
  • Cas Number : 147663-00-1
  • Molecular Weight : 185.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form grey solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-4-dihydro-1h-isochromen-4-amine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7548
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart