ADV428293202 AChemBlock

3-Iodo-1H-pyrazolo[4,3-b]pyridine

  • MDL number MFCD10696864
  • Cas Number : 633328-40-2
  • Molecular Weight : 245.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-1h-pyrazolo-4-3-b-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7560
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart