ADV428293230 AChemBlock

3-Bromo-4-chloro-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD11616444
  • Cas Number : 920966-51-4
  • Molecular Weight : 371.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-chloro-1-phenylsulfonyl-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7590
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart