ADV428293231 AChemBlock

Methyl 6-chloro-1,2,4-triazolo[4,3-b]pyridazine-3-carboxylate

  • MDL number MFCD17016121
  • Cas Number : 76196-04-8
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-chloro-1-2-4-triazolo-4-3-b-pyridazine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7591
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart