ADV428293235 AChemBlock

6-Chloro-3-nitroimidazo[1,2-b]pyridazine

  • MDL number MFCD08276991
  • Cas Number : 18087-76-8
  • Molecular Weight : 198.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3-nitroimidazo-1-2-b-pyridazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7595
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart