ADV428293237 AChemBlock

1,1-Dimethyl-2,3-dihydro-1H-isoindole

  • MDL number MFCD19687983
  • Cas Number : 712262-06-1
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-1-dimethyl-2-3-dihydro-1h-isoindole.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7597
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart