ADV428293242 AChemBlock

5-(Aminomethyl)hexahydrocyclopenta[c]pyrrole-2(1H)-carboxylic acid 1,1-dimethylethyl ester

  • MDL number MFCD23106340
  • Cas Number : 1256958-47-0
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-aminomethyl-hexahydrocyclopenta-c-pyrrole-2-1h-carboxylic-acid-1-1-dimethylethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7602
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart