ADV428293264 AChemBlock

(3S,4S)-3,4-Tetrahydrofurandiamine

  • MDL number MFCD18833628
  • Cas Number : 45434-73-9
  • Molecular Weight : 102.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-4s-3-4-tetrahydrofurandiamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7624
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart