ADV428293267 AChemBlock

1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine

  • MDL number MFCD16039627
  • Cas Number : 1254982-25-6
  • Molecular Weight : 223.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow liquid
Shipping Temperature Store at -20
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1-2-3-6-tetrahydropyridine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7627
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart