ADV428293271 AChemBlock

trans-1,4-Cyclohexanedicarboxylic acid monomethyl ester

  • MDL number MFCD01311247
  • Cas Number : 15177-67-0
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form off-white flakes
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-1-4-cyclohexanedicarboxylic-acid-monomethyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7632
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart