ADV428293276 AChemBlock

Tetrahydro-2,2,6,6-tetramethylpyran-4-one

  • MDL number MFCD12828096
  • Cas Number : 1197-66-6
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tetrahydro-2-2-6-6-tetramethylpyran-4-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7637
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart