ADV428293283 AChemBlock

(2R,6R)-tert-Butyl 2,6-dimethylpiperazine-1-carboxylate

  • MDL number MFCD11973747
  • Cas Number : 574007-62-8
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2r-6r-tert-butyl-2-6-dimethylpiperazine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7644
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart