ADV428293284 AChemBlock

(S)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride

  • MDL number MFCD23105798
  • Cas Number : 1089280-14-7
  • Molecular Weight : 215.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-octahydropyrazino-2-1-c-1-4-oxazine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7645
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart