ADV428293291 AChemBlock

2,7-Diazaspiro[4.4]nonan-1-one hydrochloride

  • MDL number MFCD10700079
  • Cas Number : 1203682-17-0
  • Molecular Weight : 176.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-7-diazaspiro-4-4-nonan-1-one-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7652
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart