ADV428293316 AChemBlock

1-Boc-3,3-difluoro-4-(hydroxymethyl)pyrrolidine

  • MDL number MFCD12755195
  • Cas Number : 1260852-42-3
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-3-difluoro-4-hydroxymethyl-pyrrolidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7679
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart