ADV428293321 AChemBlock

1-Methyl-4-{1-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]cyclopropyl}piperazine

  • MDL number MFCD27987000
  • Cas Number : 1643923-77-6
  • Molecular Weight : 342.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-4-1-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-cyclopropyl-piperazine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7684
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart