ADV428293354 AChemBlock

4-Methoxy-7-methyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD27987045
  • Cas Number : 1523606-42-9
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-7-methyl-7h-pyrrolo-2-3-d-pyrimidine-6-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7718
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart