ADV428293356 AChemBlock

1-(2-Fluoro-benzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride

  • MDL number MFCD18432772
  • Cas Number : 256499-19-1
  • Molecular Weight : 305.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-fluoro-benzyl-1h-pyrazolo-3-4-b-pyridine-3-carboxamidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-3462
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart