ADV428293359 AChemBlock

1-(Azetidin-3-yl)-3-fluoroazetidine oxalate

  • MDL number MFCD27987049
  • Cas Number : 1426290-05-2
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-azetidin-3-yl-3-fluoroazetidine-oxalate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7723
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart