ADV428293362 AChemBlock

5-Cyclopropyl-1,3,4-oxadiazole-2-carboxylic acid Lithium salt

  • MDL number MFCD28023826
  • Cas Number : 1523571-22-3
  • Molecular Weight : 160.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-cyclopropyl-1-3-4-oxadiazole-2-carboxylic-acid-lithium-salt.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7726
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart