ADV428293363 AChemBlock

trans-3-Oxabicyclo[3.1.0]hexane-6-acetic acid

  • MDL number MFCD19229296
  • Cas Number : 1110650-68-4
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/trans-3-oxabicyclo-3-1-0-hexane-6-acetic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7727
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart