ADV428293369 AChemBlock

5-(3-Fluoro-cyclobutyl)-1H-pyrazol-3-ylamine

  • MDL number MFCD27987067
  • Cas Number : 2228036-45-9
  • Molecular Weight : 155.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-3-fluoro-cyclobutyl-1h-pyrazol-3-ylamine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7733
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart