ADV428293370 AChemBlock

Methyl 6-bromo-1H-Pyrrolo[3,2-b]pyridine-2-carboxylate

  • MDL number MFCD27987068
  • Cas Number : 1352492-16-0
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromo-1h-pyrrolo-3-2-b-pyridine-2-carboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=G-7734
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart